Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-methylimidazolium Nitrate 98.0+%, TCI America™
SDP

Catalog No. E07755G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07755G 5 g
E077525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07755G Supplier TCI America Supplier No. E07755G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 143314-14-1
Molecular Formula C6H11N3O3
Molecular Weight (g/mol) 173.172
MDL Number MFCD00210007
InChI Key JDOJFSVGXRJFLL-UHFFFAOYSA-N
Synonym 1-ethyl-3-methylimidazolium nitrate, 1-ethyl-3-methylimidazolium nitrate nt, emim no3, acmc-20dtjx, dsstox_cid_29073, dsstox_rid_83292, dsstox_gsid_49217, ksc170o1h, 1-ethyl-3-methyl-1h-imidazol-3-ium nitrate
PubChem CID 16211072
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;nitrate
SMILES CCN1C=C[N+](=C1)C.[N+](=O)([O-])[O-]

Specifications

Melting Point 41°C
Color White-Yellow
Quantity 5 g
UN Number 1479
Formula Weight 173.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethyl-3-methylimidazolium Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.