Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Ethyl-3-Methylimidazolium Tetrachloroaluminate

Catalog No. 510591KGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-059-1KGF 1 kg
51-059-100GF 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 51-059-1KGF Supplier Sigma Aldrich Supplier No. 510591KGF
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Ethyl-3-Methylimidazolium Tetrachloroaluminate

Specifications

CAS 80432-05-9
Density 1.304 g/mL at 25°C
Molecular Formula C6H11AlCl4N2
Linear Formula C6H11AlCl4N2
Refractive Index n20/D 1.484
MDL Number MFCD06798187
Quantity 1 kg
Synonym 1-Ethyl-3-methyl-1H-imidazolium tetrachloroaluminate
Molecular Weight (g/mol) 279.96 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.