Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-methylimidazolium Tetrachloroferrate 98.0+%, TCI America™
SDP

Catalog No. E07065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E07065G Supplier TCI America Supplier No. E07065G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 850331-04-3
Molecular Formula C6H11Cl4FeN2
Molecular Weight (g/mol) 308.813
InChI Key VGSZFQMHQHFXCD-UHFFFAOYSA-J
PubChem CID 44629868
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;tetrachloroiron(1-)
SMILES CCN1C=C[N+](=C1)C.Cl[Fe-](Cl)(Cl)Cl

Specifications

Color Brown-Yellow
Quantity 5 g
Formula Weight 308.81
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-methylimidazolium Tetrachloroferrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.