Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Ethyl-3-Methylimidazolium Trifluoromethanesulfonate

Catalog No. 0436750G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-367-50G 50 g
04-367-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04-367-50G Supplier Sigma Aldrich Supplier No. 0436750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Ethyl-3-methylimidazolium trifluoromethanesulfonate may be used as a solvent to produce Ionic Polymer-Polymer Composites (IP2C) and also in lipase-catalyzed enantioselective amine acylation with 4-pentenoic acid.Ionic liquid stable to air and water useful for electrochemical purposes (molten salt technique) exhibiting an electrochemical stability window of Gd-4 V and being stable to up to 350 C

Specifications

CAS 145022-44-2
Density 1.387 g/mL at 25°C
Boiling Point >350°C
Molecular Formula C7H11F3N2O3S
Refractive Index n20/D 1.435; n20/D 1.435
Linear Formula C7H11F3N2O3S
MDL Number MFCD00210008
Quantity 50 g
Synonym [EMIM][TRIFLATE]; EMIM Otf
Molecular Weight (g/mol) 260.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.