Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-methylimidazolium Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. E04945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04945G 5 g
E049425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04945G Supplier TCI America Supplier No. E04945G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 145022-44-2
Molecular Formula C7H11F3N2O3S
Molecular Weight (g/mol) 260.231
MDL Number MFCD00210008
InChI Key ZPTRYWVRCNOTAS-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium trifluoromethanesulfonate, 1-ethyl-3-methylimidazolium triflate, 1-ethyl-3-methyl-1h-imidazol-3-ium trifluoromethanesulfonate, acmc-209ctv, dsstox_cid_27941, dsstox_rid_82693, dsstox_gsid_47965, 1-ethyl-3-methylimidazoliumtrifluoromethanesulfonate, ksc685i3d, 1-ethyl-3-methylimidazolium trifluoromethanesulfon
PubChem CID 2758874
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;trifluoromethanesulfonate
SMILES CCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Color Yellow
Quantity 5 g
Formula Weight 260.23
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-methylimidazolium Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.