Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-piperidone Hydrochloride 97%, Thermo Scientific™

Catalog No. AC157180250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC157180250 25 g
AC157180050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC157180250 Supplier Thermo Scientific Chemicals Supplier No. 157180250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41361-28-8
Molecular Formula C7H14ClNO
Molecular Weight (g/mol) 163.645
InChI Key JLBJGYHSFYAUDJ-UHFFFAOYSA-N
Synonym 1-ethylpiperidin-3-one hydrochloride, 1-ethyl-3-piperidone hydrochloride, 1-ethylpiperidin-3-one, chloride, 1-ethyl-3-piperidone hydrochlorid, 1-ethyl-3-piperidinone hydrochloride
PubChem CID 16211252
IUPAC Name 1-ethylpiperidin-3-one;hydrochloride
SMILES CCN1CCCC(=O)C1.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 25 g
Formula Weight 163.65
Percent Purity ≥96.0%
Physical Form Chunks and/or Powder
Chemical Name or Material 1-Ethyl-3-piperidone hydrochloride, 97%
EINECSNumber 255-333-6
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.