Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-4-(4-fluoro-2-nitrophenyl)piperazine, 97%, Thermo Scientific™

Catalog No. AAH63999MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63999MD 250 mg
AAH6399903 1 g
AAH6399906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63999MD Supplier Thermo Scientific Chemicals Supplier No. H63999MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1285896-76-5
Molecular Formula C12H16FN3O2
Molecular Weight (g/mol) 253.277
InChI Key IWKXGRRMWASTNC-UHFFFAOYSA-N
Synonym 1-ethyl-4-4-fluoro-2-nitrophenyl piperazine
PubChem CID 52438092
IUPAC Name 1-ethyl-4-(4-fluoro-2-nitrophenyl)piperazine
SMILES CCN1CCN(CC1)C2=C(C=C(C=C2)F)[N+](=O)[O-]

Specifications

Quantity 250 mg
Formula Weight 253.27
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Ethyl-4-(4-fluoro-2-nitrophenyl)piperazine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.