Learn More
Medchemexpress LLC 1-Ethyl-4-ethynylbenzene | 40307-11-7 | 98.11% | 130.19 | 25 G
SDP
Catalog No. 5000451380
Encompass_Preferred
1-Ethyl-4-ethynylbenzene is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Used as a biological material or organic compound for life science research.
  • Functions as a click chemistry reagent.
  • Contains an Alkyne group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
Catalog No. 50-004-51380 Supplier Medchemexpress LLC Supplier No. 25GHYW012717
1-Ethyl-4-ethynylbenzene is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Used as a biological material or organic compound for life science research.
  • Functions as a click chemistry reagent.
  • Contains an Alkyne group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.