Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Ethylpyridinium Tetrafluoroborate

Catalog No. 7131045G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-310-45G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-310-45G Supplier Sigma Aldrich Supplier No. 7131045G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Ethylpyridinium tetrafluoroborate [EPy][BF4} is an ionic liquid that can be used as:A reaction medium in the Morita-Baylis-Hillman (MBH) reactions of acrylonitrile with 4-nitrobenzaldehyde to yield MBH adducts.A solvent in the Diels-Alder reactionsof isoprenewith dienophiles.A component of a binary mixture, which is employed in the solute-solvent interaction studies using vitamins like nicotinic acid and ascorbic acid.

Specifications

Molecular Formula C7H16BF4N
Linear Formula C7H16BF4N
MDL Number MFCD08458505
Quantity 50 g
Molecular Weight (g/mol) 201.01 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.