Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Ethynyl-1-cyclohexanol

Catalog No. E51406100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 L
5 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E51406100ML 100 mL
E514061L 1 L
E5140620L 20 L
E514065L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. E51406100ML Supplier Sigma Aldrich Supplier No. E51406100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Ethynyl-1-cyclohexanol

Specifications

CAS 78-27-3
Melting Point 30°C to 33°C (lit.)
Density 0.967 g/mL (at 25°C (literature))
Boiling Point 180°C (lit.)
Molecular Formula C8H12O
Linear Formula HC///CC6H10OH
MDL Number MFCD00003858
Quantity 100 mL
Molecular Weight (g/mol) 124.18
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.