Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethynyl-1-cyclohexanol, 99+%

Catalog No. AC119101000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC119101000 100 g Glass bottle
AC119105000 500 g Glass bottle
AC119100025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC119101000 Supplier Thermo Scientific Chemicals Supplier No. 119101000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 78-27-3 | C8H12O | 124.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-27-3
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.18
MDL Number MFCD00003858
InChI Key QYLFHLNFIHBCPR-UHFFFAOYSA-N
Synonym 1-ethynyl-1-cyclohexanol, 1-ethynylcyclohexanol, ethynylcyclohexanol, cyclohexanol, 1-ethynyl, 1-hydroxycyclohexyl ethyne, 1-ethynyl-1-hydroxycyclohexane, 1-ethynyl cyclohexanol, 1-ethynyl-cyclohexanol, unii-6rv04025eh, 1-ethynyl-cyclohexan-1-ol
PubChem CID 6525
IUPAC Name 1-ethynylcyclohexan-1-ol
SMILES C#CC1(CCCCC1)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 30.0°C to 32.0°C
Color Colorless to Yellow
Density 0.9763g/mL
Boiling Point 180.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4810 to 1.4840
Linear Formula HC≡CC6H10OH
Quantity 100 g
Beilstein 06,II,1
Specific Gravity 0.9763
Merck Index 10,381
Solubility Information Solubility in water: 24.5g/L (20°C). Other solubilities: soluble in most organic solvents, soluble in diethyl ether, alcohols, esters, ketone, and chlorinated hydrocarbons
Formula Weight 124.18
Percent Purity 99+%
Physical Form Liquid After Melting
Chemical Name or Material 1-Ethynyl-1-cyclohexanol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Toxic in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
Take off contaminated clothing a
EINECSNumber 201-100-9
RTECSNumber GV9100000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.