Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethynylcyclohexylamine, 98%

Catalog No. p-7035787
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5351406 5 g
AAH5351414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5351406 Supplier Thermo Scientific Chemicals Supplier No. H5351406
Only null left
Add to Cart

CAS: 30389-18-5 | C8H13N | 123.20 g/mol

1-Ethynylcyclohexylamine was used in the synthesis of bicyclic and tetracyclic compounds. It was also used as a substrate in the reaction of a series of alkynes with mercury(II)acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethynylcyclohexylamine was used in the synthesis of bicyclic and tetracyclic compounds. It was also used as a substrate in the reaction of a series of alkynes with mercury(II)acetate.

Notes
Air sensitive. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30389-18-5
Molecular Formula C8H13N
Molecular Weight (g/mol) 123.20
MDL Number MFCD00001489
InChI Key GDKOYYDQISQOMH-UHFFFAOYSA-N
PubChem CID 121691
IUPAC Name 1-ethynylcyclohexan-1-amine
SMILES NC1(CCCCC1)C#C

Specifications

Density 0.913
Boiling Point 65°C to 66°C (20 mmHg)
Flash Point 42°C (108°F)
Refractive Index 1.482
Quantity 5 g
UN Number UN2734
Sensitivity Air Sensitive
Synonym 1-ethynylcyclohexylamine, 1-ethynylcyclohexanamine, cyclohexanamine, 1-ethynyl, cyclohexanamine,1-ethynyl, 1-ethynyl-cyclohexylamine, 1-ethynylcyclohexaneamine, 1-ethynylcyclohexyl amine, 1-ethinyl-cyclohexyl-amine, 1-ethynyl-1-amino-cyclohexane
Formula Weight 123.2
Percent Purity 98%
Chemical Name or Material 1-Ethynylcyclohexylamine
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 250-172-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.