Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fluorenecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. F05411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05411G 1 g
F05415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05411G Supplier TCI America Supplier No. F05411G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6276-03-5
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.23
MDL Number MFCD00001130
InChI Key HTPXFGUCAUTOEL-UHFFFAOYSA-N
Synonym 1-fluorenecarboxylic acid, fluorene-1-carboxylic acid, fluorenecarboxylic acid, unii-ao792vd660, 1-fluorene carboxylic acid, 9h-fluorene-1-carboxylicacid, htpxfgucautoel-uhfffaoysa
PubChem CID 80468
IUPAC Name 9H-fluorene-1-carboxylic acid
SMILES C1C2=CC=CC=C2C3=C1C(=CC=C3)C(=O)O

Specifications

Melting Point 248°C
Color Yellow
Boiling Point 100°C
Quantity 1 g
Formula Weight 210.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Fluorenecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.