Learn More
Accel Pharmtech Inc 1-FLUORO-2,4,6-TRIMETHYLP100G
SDP
Supplier:  Accel Pharmtech Inc XK215648100G
Encompass
1-FLUORO-2,4,6-TRIMETHYLPYRIDIN-1-IUM TETRAFLUOROBORATE | Purity: 98% | Mol Wt: 226.983 | 109705-14-8 | MFCD00153182 | 100 g
Catalog No. NC2021313