Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fluoro-2,4,6-trimethylpyridinium Trifluoromethanesulfonate 95.0+%, TCI America™
SDP

Catalog No. F032825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F032825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F032825G Supplier TCI America Supplier No. F032825G

missing translation for 'validMainframeMsg'

Specifications

CAS 107264-00-6
Melting Point 165°C
Color White-Yellow
Molecular Formula C9H11F4NO3S
MDL Number MFCD00067525
UN Number 1759
Quantity 25 g
Synonym 1-fluoro-2,4,6-trimethylpyridinium triflate, n-fluoro-2,4,6-trimethylpyridinium triflate, 1-fluoro-2,4,6-trimethylpyridin-1-ium trifluoromethanesulfonate, 1-fluoro-2,4,6-trimethylpyridin-1-ium triflate, 1-fluoro-2,4,6-trimethylpyridinium trifluoromethanesulfonate, 1-fluoro-2,4,6-trimethylpyridin-1-ium; trifluoromethanesulfonate, 1-fluoro-2,4,6-trimethylpyridine, trifluoromethanesulfonic acid, acmc-1bprl, 1-fluoro-2,4,6-trimethylpyridiniumtriflate, 1-fluoro-2,4,6-trimethyl-pyridinium triflate
InChI Key PRIGFEJKMMRJSF-UHFFFAOYSA-M
SMILES CC1=CC(=[N+](C(=C1)C)F)C.C(F)(F)(F)S(=O)(=O)[O-]
IUPAC Name 1-fluoro-2,4,6-trimethylpyridin-1-ium;trifluoromethanesulfonate
Molecular Weight (g/mol) 289.245
PubChem CID 2724575
Formula Weight 289.24
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Fluoro-2,4,6-trimethylpyridinium Trifluoromethanesulfonate [Fluorinating Reagent]
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.