Learn More
Sigma Aldrich Fine Chemicals Biosciences 1-Fluoro-2,4-dinitrobenzene for HPLC derivatization, LiChropur(TM), >=99.0% (GC) | 70-34-8 | MFCD00007056 | 1G
Catalog No. 501840517
Encompass
1-Fluoro-2,4-dinitrobenzene for HPLC derivatization, LiChropur(TM), >=99.0% (GC) | Purity: >=99.0% (GC) | Mol Wt: 186.1 | 70-34-8 | MFCD00007056 | 1G
Catalog No. 501840517 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 731771G
1-Fluoro-2,4-dinitrobenzene for HPLC derivatization, LiChropur(TM), >=99.0% (GC) | Purity: >=99.0% (GC) | Mol Wt: 186.1 | 70-34-8 | MFCD00007056 | 1G