Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Fluoro-2-nitrobenzene

Catalog No. F10801250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10801250G 250 g
F1080150G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10801250G Supplier Sigma Aldrich Supplier No. F10801250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Fluoro-2-nitrobenzene

Specifications

CAS 1493-27-2
Melting Point -9°C to +6°C (lit.)
Density 1.338 g/mL (at 25°C (literature))
Boiling Point 116°C (22 mmHg)
Molecular Formula C6H4FNO2
Refractive Index n20/D 1.532 (literature)
Linear Formula FC6H4NO2
MDL Number MFCD00007048
Quantity 250 g
Molecular Weight (g/mol) 141.1
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.