1-Fluoro-2-nitrobenzene, 99%

Manufacturer: Alfa Aesar A1385218

Quantity 50g

 View more versions of this product

Catalog No. AAA1385218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1493-27-2
    Molecular Formula C6H4FNO2
    Formula Weight 141.1
    MDL Number MFCD00007048
    UN Number UN2810
    Beilstein 607262
    Chemical Name or Material 1-Fluoro-2-nitrobenzene

    Specifications

    Formula Weight 141.1
    Quantity 50g
    TSCA TSCA
    MDL Number MFCD00007048
    UN Number UN2810
    Beilstein 607262
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H373-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    Causes serious ey
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several mi
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Physical Form Liquid
    Density 1.340
    Assay Percent Range 99%
    Molecular Formula C6H4FNO2
    CAS 1493-27-2
    Chemical Name or Material 1-Fluoro-2-nitrobenzene
    Assay 99%

    Description

    Applications
    1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.

    Solubility
    Immiscible in water.

    Notes
    Avoid excessive heat and light.

    GHS H Statement
    H301-H311-H331-H373-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    Causes serious ey

    GHS P Statement
    P260-P301+P310-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several mi

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit