Learn More
Sigma Aldrich Fine Chemicals Biosciences 1 Fluoro 24 dinitrobenzene
Catalog No. 502906991
Encompass
1-Fluoro-2 4-dinitrobenzene (DNFB) is a reagent that modifies the N-terminal amino acids of proteins and peptides. DNFB is also commonly known as Sangerprimes Reagent named after Frederick Sanger who reported in a classic 1945 paper the use of DNFB for derivatization of proteins and peptides specifically at their N-termini.x2020
Catalog No. 50-290-6991 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D1529100ML
1-Fluoro-2 4-dinitrobenzene (DNFB) is a reagent that modifies the N-terminal amino acids of proteins and peptides. DNFB is also commonly known as Sangerprimes Reagent named after Frederick Sanger who reported in a classic 1945 paper the use of DNFB for derivatization of proteins and peptides specifically at their N-termini.x2020