Learn More
Medchemexpress LLC 1-fluoro-3-(2-(4-(methylamino)phenyl)quinolin-6-yloxy)propan-2-ol | 1374107-54-6 | 98.5% | 326.36 g/mol | C19H19FN2O2 | 10 MG
SDP
Catalog No. 5000213168
Encompass_Preferred
THK-5117 is an arylquinoline derivative research compound that binds tau fibrils and is used in biochemical and preclinical studies of tau pathology. Supplied as a solid with characterized molecular properties, it is intended for laboratory research use only.
  • High binding affinity to tau fibrils (Ki ≈ 10.5 nM).
  • High purity (≈98.5%) for reproducible results.
  • Molecular weight 326.36 g/mol for accurate dosing calculations.
  • Supplied as a solid suitable for standard laboratory handling.
  • Includes datasheet, certificate of analysis, and safety data sheet.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-13168 Supplier Medchemexpress LLC Supplier No. HY10118210MG
THK-5117 is an arylquinoline derivative research compound that binds tau fibrils and is used in biochemical and preclinical studies of tau pathology. Supplied as a solid with characterized molecular properties, it is intended for laboratory research use only.
  • High binding affinity to tau fibrils (Ki ≈ 10.5 nM).
  • High purity (≈98.5%) for reproducible results.
  • Molecular weight 326.36 g/mol for accurate dosing calculations.
  • Supplied as a solid suitable for standard laboratory handling.
  • Includes datasheet, certificate of analysis, and safety data sheet.
  • Intended for research use only; not for human therapeutic use.