Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fluoro-4-nitrobenzene, 99%

Catalog No. p-7081189
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1105722 100 g
AAA1105736 500 g
AAA110570E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1105722 Supplier Thermo Scientific Chemicals Supplier No. A1105722
Only null left
Add to Cart

CAS: 350-46-9 | C6H4FNO2 | 141.101 g/mol

1-Fluoro-4-nitrobenzene used as a component of hair dyes, as pharmaceutical intermediates. It is also used for the preparation of novel soluble aromatic polyimides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoro-4-nitrobenzene used as a component of hair dyes, as pharmaceutical intermediates. It is also used for the preparation of novel soluble aromatic polyimides

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents, acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 350-46-9
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.101
MDL Number MFCD00007282
InChI Key WFQDTOYDVUWQMS-UHFFFAOYSA-N
Synonym 4-fluoronitrobenzene, p-fluoronitrobenzene, p-nitrofluorobenzene, benzene, 1-fluoro-4-nitro, 4-nitrofluorobenzene, 1-nitro-4-fluorobenzene, 4-fluoro-1-nitrobenzene, 1-fluoro-4-nitro-benzene, unii-a2m2fh7xhh, p-fluoro nitrobenzene
PubChem CID 9590
IUPAC Name 1-fluoro-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])F

Specifications

Melting Point 22°C to 24°C
Density 1.33
Boiling Point 204°C to 206°C
Flash Point 83°C (181°F)
Refractive Index 1.5315
Quantity 100 g
UN Number UN2811
Beilstein 606923
Solubility Information Insoluble in water.
Formula Weight 141.1
Percent Purity 99%
Chemical Name or Material 1-Fluoro-4-nitrobenzene
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H331-H335-H373
Hazard Statement GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 206-502-8
RTECSNumber DA1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.