Learn More
Medchemexpress LLC 1-Fluoronaphthalene | 321-38-0 | 99.6% | 146.16 | 100 G
SDP
Catalog No. 5000400655
Encompass_Preferred
1-Fluoronaphthalene is an organofluorine compound derived from naphthalene derivatives and fluorinated aromatics. It serves as a precursor for synthesizing LY248686, a potent inhibitor of serotonin and noradrenaline uptake, making it valuable for research applications.
  • Can be used to synthesize LY248686.
  • Suitable for research applications.
  • Oxidized by *C. elegans* to various hydroxylated and conjugated forms.
  • Available in liquid form.
Catalog No. 50-004-00655 Supplier Medchemexpress LLC Supplier No. HY32126100G
1-Fluoronaphthalene is an organofluorine compound derived from naphthalene derivatives and fluorinated aromatics. It serves as a precursor for synthesizing LY248686, a potent inhibitor of serotonin and noradrenaline uptake, making it valuable for research applications.
  • Can be used to synthesize LY248686.
  • Suitable for research applications.
  • Oxidized by *C. elegans* to various hydroxylated and conjugated forms.
  • Available in liquid form.