Learn More
Medchemexpress LLC 1-Fluoronaphthalene | 321-38-0 | MFCD00003873 | 96.9% | 146.16 | 250 MG
SDP
Catalog No. 5000402919
Encompass_Preferred
1-Fluoronaphthalene (Standard) is an organofluorine compound derived from naphthalene derivatives and fluorinated aromatics. It is intended for research and analytical applications, serving as the analytical standard for 1-Fluoronaphthalene. This compound can be used to synthesize LY248686, a potent inhibitor of serotonin and noradrenaline uptake. It is commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for 1-Fluoronaphthalene.
  • Used in research and analytical applications.
  • Can be used to synthesize LY248686.
  • Suitable for HPLC, GC, and MS research experiments.
Catalog No. 50-004-02919 Supplier Medchemexpress LLC Supplier No. 250MGHY32126R
1-Fluoronaphthalene (Standard) is an organofluorine compound derived from naphthalene derivatives and fluorinated aromatics. It is intended for research and analytical applications, serving as the analytical standard for 1-Fluoronaphthalene. This compound can be used to synthesize LY248686, a potent inhibitor of serotonin and noradrenaline uptake. It is commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for 1-Fluoronaphthalene.
  • Used in research and analytical applications.
  • Can be used to synthesize LY248686.
  • Suitable for HPLC, GC, and MS research experiments.