Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fluoronaphthalene Solution, 1000μg/mL in methylene chloride

Catalog No. USIST1801
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-IST-1801 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-IST-1801 Supplier Agilent Technologies Supplier No. IST1801
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 321-38-0 , 75-09-2
Molecular Formula C10H7F
Molecular Weight (g/mol) 146.164
InChI Key CWLKTJOTWITYSI-UHFFFAOYSA-N
PubChem CID 9450
IUPAC Name 1-fluoronaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2F

Specifications

CAS Max % 99.93, 0.07
Analyte or Component Names Methylene chloride,1-fluoronaphthalene
Element Methylene Chloride, 1-fluoronaphthalene
Solvent or Matrix Methylene Chloride
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material 1-Fluoronaphthalene Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.