Learn More
Medchemexpress LLC 1-Fluoronaphthalene (standard) | 321-38-0 | 96.9% | 146.16 | 50 MG
SDP
Catalog No. 5000401454
Encompass_Preferred
1-Fluoronaphthalene (Standard) is the analytical standard of 1-Fluoronaphthalene, intended for research and analytical applications. It is an organofluorine compound derived from naphthalene derivatives and fluorinated aromatics. This compound can be used to synthesize LY248686, a potent inhibitor of serotonin and noradrenaline uptake.
  • Used as an analytical standard.
  • Commonly employed in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-01454 Supplier Medchemexpress LLC Supplier No. HY32126R50MG
1-Fluoronaphthalene (Standard) is the analytical standard of 1-Fluoronaphthalene, intended for research and analytical applications. It is an organofluorine compound derived from naphthalene derivatives and fluorinated aromatics. This compound can be used to synthesize LY248686, a potent inhibitor of serotonin and noradrenaline uptake.
  • Used as an analytical standard.
  • Commonly employed in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.