Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fluoropyridinium Trifluoromethanesulfonate 96.0+%, TCI America™
SDP

Catalog No. F03275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03275G 5 g
F032725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03275G Supplier TCI America Supplier No. F03275G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 107263-95-6
Molecular Formula C6H5F4NO3S
Molecular Weight (g/mol) 247.164
MDL Number MFCD00013458
InChI Key JFZMMCYRTJBQQI-UHFFFAOYSA-M
Synonym 1-fluoropyridinium triflate, n-fluoropyridinium triflate, n-fluoropyridinium trifluoromethanesulfonate, 1-fluoropyridinium trifluoromethanesulfonate, 1-fluoropyridin-1-ium; trifluoromethanesulfonate, acmc-209shu, ksc182s3j, 1-fluoropyridin-1-ium triflate, 1-fluoropyridiniumtriflate
PubChem CID 2724576
IUPAC Name 1-fluoropyridin-1-ium;trifluoromethanesulfonate
SMILES C1=CC=[N+](C=C1)F.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 183°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 247.16
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Fluoropyridinium Trifluoromethanesulfonate [Fluorinating Reagent]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.