Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 1-Fmoc-2-piperidineacetic acid | 10 g | CAS 683220-37-3 | MDL MFCD02667642

Catalog No. 502349548
Encompass

1-Fmoc-2-piperidineacetic acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 683220-37-3
- MDL: MFCD02667642
- InChIKey: YVHXTQQJFAOBCJ-UHFFFAOYSA-N
- Molecular Weight: 365.429
- Molecular Formula: C22H23NO4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29333999
- Country Of Origin: China
- IUPAC: 2-(1-(((9H-fluoren-9-yl)methoxy)carbonyl)piperidin-2-yl)acetic acid
- SMILES: O=C(CC1CCCCN1C(OCC2C3=CC=CC=C3C4=C2C=CC=C4)=O)O

Catalog No. 50-234-9548 Supplier STA Pharmaceutical Supplier No. STAAA1031310GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Fmoc-2-piperidineacetic acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 683220-37-3
- MDL: MFCD02667642
- InChIKey: YVHXTQQJFAOBCJ-UHFFFAOYSA-N
- Molecular Weight: 365.429
- Molecular Formula: C22H23NO4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29333999
- Country Of Origin: China
- IUPAC: 2-(1-(((9H-fluoren-9-yl)methoxy)carbonyl)piperidin-2-yl)acetic acid
- SMILES: O=C(CC1CCCCN1C(OCC2C3=CC=CC=C3C4=C2C=CC=C4)=O)O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.