Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 1-Fmoc-4-(4-fluorophenyl)-4-carboxypiperidine | 25 g | CAS 1076197-06-2 | MDL MFCD11226817

Catalog No. 502352890
Encompass

1-Fmoc-4-(4-fluorophenyl)-4-carboxypiperidine is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1076197-06-2
- MDL: MFCD11226817
- InChIKey: DPGFVVPTWGNEHH-UHFFFAOYSA-N
- Molecular Weight: 445.49
- Molecular Formula: C27H24FNO4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29333999
- Country Of Origin: China
- IUPAC: 1-(((9H-fluoren-9-yl)methoxy)carbonyl)-4-(4-fluorophenyl)piperidine-4-carboxylic acid
- SMILES: O=C(N1CCC(C2=CC=C(C=C2)F)(CC1)C(O)=O)OCC3C4=CC=CC=C4C5=CC=CC=C35

Catalog No. 50-235-2890 Supplier STA Pharmaceutical Supplier No. STAAA1031225GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Fmoc-4-(4-fluorophenyl)-4-carboxypiperidine is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1076197-06-2
- MDL: MFCD11226817
- InChIKey: DPGFVVPTWGNEHH-UHFFFAOYSA-N
- Molecular Weight: 445.49
- Molecular Formula: C27H24FNO4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29333999
- Country Of Origin: China
- IUPAC: 1-(((9H-fluoren-9-yl)methoxy)carbonyl)-4-(4-fluorophenyl)piperidine-4-carboxylic acid
- SMILES: O=C(N1CCC(C2=CC=C(C=C2)F)(CC1)C(O)=O)OCC3C4=CC=CC=C4C5=CC=CC=C35

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.