Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 1-Fmoc-4-(hydroxymethyl)piperidine | 5 g | CAS 1072502-03-4 | InChIKey YYTSXYVXBGQKCP-UHFFFAOYSA-N

Catalog No. 502341546
Encompass

1-Fmoc-4-(hydroxymethyl)piperidine is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1072502-03-4
- MDL: No data
- InChIKey: YYTSXYVXBGQKCP-UHFFFAOYSA-N
- Molecular Weight: 337.419
- Molecular Formula: C21H23NO3
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29333999
- Country Of Origin: China
- IUPAC: (9H-fluoren-9-yl)methyl 4-(hydroxymethyl)piperidine-1-carboxylate
- SMILES: OCC1CCN(C(OCC2C3=C(C4=C2C=CC=C4)C=CC=C3)=O)CC1

Catalog No. 50-234-1546 Supplier STA Pharmaceutical Supplier No. STAAA117375GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Fmoc-4-(hydroxymethyl)piperidine is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1072502-03-4
- MDL: No data
- InChIKey: YYTSXYVXBGQKCP-UHFFFAOYSA-N
- Molecular Weight: 337.419
- Molecular Formula: C21H23NO3
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29333999
- Country Of Origin: China
- IUPAC: (9H-fluoren-9-yl)methyl 4-(hydroxymethyl)piperidine-1-carboxylate
- SMILES: OCC1CCN(C(OCC2C3=C(C4=C2C=CC=C4)C=CC=C3)=O)CC1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.