Learn More
Medchemexpress LLC 1-formyl-beta-carboline | 20127-63-3 | MFCD14706724 | 99.7% | 196.21 g/mol | C12H8N2O | 5 MG
SDP
Catalog No. 5000283358
Encompass_Preferred
1-formyl-beta-carboline is a small-molecule beta-carboline provided for research use as a biochemical probe. It has reported activity as an Akt pathway inhibitor and is used in antiviral and PI3K/Akt/mTOR pathway studies. The compound is supplied as a solid with high reported purity and available in multiple mg-scale packages for laboratory experiments.
  • High purity suitable for biochemical research.
  • Reported inhibition of Akt signaling pathways.
  • Useful for antiviral and pathway modulation assays.
  • Available in multiple mg-scale package sizes.
  • Supplied with data sheet and certificate of analysis for characterization.
Catalog No. 50-002-83358 Supplier Medchemexpress LLC Supplier No. HYW1309655MG
1-formyl-beta-carboline is a small-molecule beta-carboline provided for research use as a biochemical probe. It has reported activity as an Akt pathway inhibitor and is used in antiviral and PI3K/Akt/mTOR pathway studies. The compound is supplied as a solid with high reported purity and available in multiple mg-scale packages for laboratory experiments.
  • High purity suitable for biochemical research.
  • Reported inhibition of Akt signaling pathways.
  • Useful for antiviral and pathway modulation assays.
  • Available in multiple mg-scale package sizes.
  • Supplied with data sheet and certificate of analysis for characterization.