Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Formylcyclopropyl)carbamic acid tert-butyl ester, 97%, Thermo Scientific™

Catalog No. AC449950010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449950010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC449950010 Supplier Thermo Scientific Chemicals Supplier No. 449950010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 107259-06-3
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.22
InChI Key ACMHCEYCNRURST-UHFFFAOYSA-N
Synonym tert-butyl 1-formylcyclopropyl carbamate, tert-butyl 1-formylcyclopropylcarbamate, 1-formyl-cyclopropyl-carbamic acid tert-butyl ester, tert-butyl n-1-formylcyclopropyl carbamate, pubchem17629, tert-butyl 1-formylcyclopropyl-carbamate, n-boc-1-amino-1-cyclopropanecarboxaldehyde, tert-butyl l-formylcyclopropyl-carbamate
PubChem CID 10352385
IUPAC Name tert-butyl N-(1-formylcyclopropyl)carbamate
SMILES CC(C)(C)OC(=O)NC1(CC1)C=O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 1 g
Formula Weight 185.22
Percent Purity 97%
Chemical Name or Material (1-Formylcyclopropyl)carbamic acid tert-butyl ester
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid releas

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.