Learn More
Newcomer Supply Phosphate Buffer 0.1M, pH 7.2 to 7.4, 1 gallon
SDP
Catalog No. NC1782639
Encompass
Use: IHC wash and rinse. Shelf Life is 1 year from date of manufacture. Store at 2-8°C.
Catalog No. NC1782639 Supplier Newcomer Supply Supplier No. 13313B
Use: IHC wash and rinse. Shelf Life is 1 year from date of manufacture. Store at 2-8°C.