Learn More
US Plastic Corporation 1 GAL ECO BLK RND BUCKET
SDP
Catalog No. NC2102593
Encompass
1 Gallon Economy Black Round HDPE Bucket with Wire Bail. BPA free. 75 mil thickness. Dimensions: 8" Top OD x 7" Bottom OD x 7-7/8" Hgt. Lids sold separately.
This item is not returnable. View return policy
Catalog No. NC2102593 Supplier US Plastic Corporation Supplier No. 18100
1 Gallon Economy Black Round HDPE Bucket with Wire Bail. BPA free. 75 mil thickness. Dimensions: 8" Top OD x 7" Bottom OD x 7-7/8" Hgt. Lids sold separately.