Learn More
US Plastic Corporation 1 Gallon Wide Mouth Round Natural HDPE Jar with White Ribbed Cap
SDP
Catalog No. NC9855465
Encompass
Natural high density polyethylene plastic with high corrosion resistance. Much lighter than glass jars. For various uses such as samples, storage of liquids, pastes and powders. White plastic caps included. Meets FDA standards.
Catalog No. NC9855465 Supplier US Plastic Corporation Supplier No. 66030
Natural high density polyethylene plastic with high corrosion resistance. Much lighter than glass jars. For various uses such as samples, storage of liquids, pastes and powders. White plastic caps included. Meets FDA standards.