Learn More
Andwin Scientific 1 GALLON BACK UP WASTE CONTAINER
SDP
Catalog No. NC3802102
Encompass_Preferred
Carlisle PS802 1 GALLON BACK UP WASTE CONTAINER CS/6 · Made of high density, break-resistant polyethylene· Easy clean containers have straight wall design with no sharp interior edges that can collect food or bacteria· Color-coordinated spouts and caps for easy product identification· Prepare mixes and juices in advance· Backup unit includes cap and container· dishwasher safe· BPA free· Assorted packs include 1 each of following colors: red, green, yellow, orange, brown and white· Exclusive manufacturing process delivers strong leak resistant seal· Economical polyethylene will withstand temperatures from -40° to 180° F
Catalog No. NC3802102 Supplier Andwin Scientific Supplier No. JPLPS802
Carlisle PS802 1 GALLON BACK UP WASTE CONTAINER CS/6 · Made of high density, break-resistant polyethylene· Easy clean containers have straight wall design with no sharp interior edges that can collect food or bacteria· Color-coordinated spouts and caps for easy product identification· Prepare mixes and juices in advance· Backup unit includes cap and container· dishwasher safe· BPA free· Assorted packs include 1 each of following colors: red, green, yellow, orange, brown and white· Exclusive manufacturing process delivers strong leak resistant seal· Economical polyethylene will withstand temperatures from -40° to 180° F