Learn More
Cole-Parmer Stackable high-density polyethylene pail, 1 gallon
Supplier:  Cole-Parmer 0627400
Encompass_Preferred
Pails conform to USDA specifications and are FDA compliant. Tapered body permits one pail to nest inside another for easy storage and shipping. Pails have bail-type handle and recessed fingergrip rim. Cover features a ''tear-tab'' strip that allows easy access to product; no lid opener required. Cover is not included.
Catalog No. NC1567513