Learn More
Uline 1 GALLON PLASTIC PAIL, WHITE
Catalog No. NC0368071
Encompass_Preferred
This item has a minimum order quantity of 5 per supplier requirements. High-density polyethylene construction withstands temperatures up to 180 degrees F. Stackable but can easily separate due to tapered design.
Catalog No. NC0368071 Supplier Uline Supplier No. S17941W
This item has a minimum order quantity of 5 per supplier requirements. High-density polyethylene construction withstands temperatures up to 180 degrees F. Stackable but can easily separate due to tapered design.