Learn More
TELEDYNE LECROY INC 1 GHZ 4 16 CH 12 BITS 10 GS
Catalog No. NC3835804
Encompass_Preferred
We’re currently building out the product information and will be adding additional content soon. Thank you for your patience as we continue to expand our selection and improve your ordering experience.
Catalog No. NC3835804 Supplier TELEDYNE LECROY INC Supplier No. HDO6104BMS
Add to Cart
Add to Cart
Request a Quote