Learn More
Hach Company 1 Glass 2.5gal Bottle SD900
Supplier:  Hach Company RF010025
Encompass
Kit-1 Glass 2 5 gallon Bottle for SD900 Refrigerated SamplerRefrigerated bottle kit with (1) 2 5 gal glass bottle with cap and full bottle shut-off
Catalog No. 50-002-07155