Learn More
Hach Company 1 Glass 2.5gal Bottle SD900
Supplier:  Hach Company PS010025
Encompass
Kit-1 Bottle-2 5 Gallon-Glass-for SD900 Portable Standard SamplerStandard Portable Bottle Kit with (1) 2 5 gal glass bottle with cap and full bottle shut off
Catalog No. 50-002-07061