Learn More
TA Instruments 1 Gram Class E2 Precision Weight, Weight Calibration, TGA Instruments, 1 Gram Class E2 Precision Weight
Catalog No. 5000580819 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments 1 gram Class E2 Precision Weight is used for weight calibration of TGA instruments. It allows users to maintain accurate weight measurements for reliable analytical results.
Catalog No. 50-005-80819 Supplier TA Instruments Supplier No. 269931.002
Waters TA Instruments 1 gram Class E2 Precision Weight is used for weight calibration of TGA instruments. It allows users to maintain accurate weight measurements for reliable analytical results.