Learn More
Berry & Associates Inc Molecular Trap, 1 gram, activated molecular sieve packet, for solvent dehydration, maintains <50 ppm water levels, prevents valve clogging, 60 mL recommended solvent volume
Catalog No. NC1327142
Encompass
Molecular Trap, 1 gram, activated molecular sieve packet, for solvent dehydration, maintains <50 ppm water levels, prevents valve clogging, 60 mL recommended solvent volume
Catalog No. NC1327142 Supplier Berry & Associates Inc Supplier No. BG700121
Molecular Trap, 1 gram, activated molecular sieve packet, for solvent dehydration, maintains <50 ppm water levels, prevents valve clogging, 60 mL recommended solvent volume