Learn More
Medchemexpress LLC 1αH,5αH-Guaia-6-ene-4β,10β-diol | 2013537-81-8 | 5 MG
SDP
Catalog No. 5000418537
Encompass_Preferred
1αH,5αH-Guaia-6-ene-4β,10β-diol is a sesquiterpenoid derivative identified from *Alisma orientale*. This compound has demonstrated anti-cancer activities.
  • Sesquiterpenoid derivative identified from *Alisma orientale*
  • Has anti-cancer activities
  • Active against H460, MCF-7, and PC-7 cell lines with IC50 values of 56.7 μM, 76.7 μM, and 59.0 μM, respectively
Catalog No. 50-004-18537 Supplier Medchemexpress LLC Supplier No. HYN50815MG
1αH,5αH-Guaia-6-ene-4β,10β-diol is a sesquiterpenoid derivative identified from *Alisma orientale*. This compound has demonstrated anti-cancer activities.
  • Sesquiterpenoid derivative identified from *Alisma orientale*
  • Has anti-cancer activities
  • Active against H460, MCF-7, and PC-7 cell lines with IC50 values of 56.7 μM, 76.7 μM, and 59.0 μM, respectively