Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1αH,5αH-guaia-6-ene-4β,10β-diol | 2013537-81-8 | 96.7% | 238.37 g·mol-1 | C15H26O2 | 10 MG
SDP

Catalog No. 5000289817
Encompass_Preferred

1αH,5αH-guaia-6-ene-4β,10β-diol is a sesquiterpenoid derivative identified from Alisma orientale with reported anti-cancer activity in cell lines. Supplied for research use in milligram quantities.

  • Sesquiterpenoid small molecule with reported anti-cancer activity.
  • Isolated from Alisma orientale.
  • Molecular formula C15H26O2; molecular weight 238.37 g·mol-1.
  • CAS number 2013537-81-8.
  • Purity listed as 96.72% by supplier.
  • Available in 5 mg, 10 mg, 50 mg, and 100 mg quantities.
  • For research use only; not for human use.

Catalog No. 50-002-89817 Supplier Medchemexpress LLC Supplier No. HYN508110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1αH,5αH-guaia-6-ene-4β,10β-diol is a sesquiterpenoid derivative identified from Alisma orientale with reported anti-cancer activity in cell lines. Supplied for research use in milligram quantities.

  • Sesquiterpenoid small molecule with reported anti-cancer activity.
  • Isolated from Alisma orientale.
  • Molecular formula C15H26O2; molecular weight 238.37 g·mol-1.
  • CAS number 2013537-81-8.
  • Purity listed as 96.72% by supplier.
  • Available in 5 mg, 10 mg, 50 mg, and 100 mg quantities.
  • For research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.