Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Heneicosanol 98.0+%, TCI America™
SDP

Catalog No. H001110G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H001110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H001110G Supplier TCI America Supplier No. H001110G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15594-90-8
Molecular Formula C21H44O
Molecular Weight (g/mol) 312.58
MDL Number MFCD00062834
InChI Key FIPPFBHCBUDBRR-UHFFFAOYSA-N
Synonym 1-heneicosanol, heneicosanol, henicosanol, 1-henicosanol, heneicosyl alcohol, heneicosan-1-ol, n-heneicosanol, n-henicosanol, heneicosylalkohol, henicosyl alcohol
PubChem CID 85014
ChEBI CHEBI:78410
IUPAC Name henicosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 69°C
Color White
Quantity 10 g
Formula Weight 312.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Heneicosanol
EINECSNumber (2)-0217
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.