Learn More
Medchemexpress LLC 1-heptadecanoyl-2-hydroxy-sn-glycero-3-phosphocholine | 50930-23-9 | MFCD00133433 | 99.0% | C25H52NO7P | 10MG
SDP
Catalog No. 5000260236
Encompass_Preferred
1-Heptadecanoyl-2-hydroxy-sn-glycero-3-phosphocholine (LPC 17:0) is a lysophosphatidylcholine with a heptadecanoyl (17:0) acyl chain. It is an endogenous metabolite found in blood and is supplied as a high-purity research standard for metabolomics, lipidomics, and biomarker studies, including pregnancy and sepsis research.
  • High-purity reference standard (99.0%).
  • Molecular formula C25H52NO7P; molecular weight 509.66.
  • Solid powder form for storage and handling.
  • Stable as a powder at -20°C for up to 3 years.
  • Suitable for metabolomics and lipidomics assays.
  • Commonly used as an endogenous biomarker in pregnancy and sepsis research.
Catalog No. 50-002-60236 Supplier Medchemexpress LLC Supplier No. HY14161610MG
1-Heptadecanoyl-2-hydroxy-sn-glycero-3-phosphocholine (LPC 17:0) is a lysophosphatidylcholine with a heptadecanoyl (17:0) acyl chain. It is an endogenous metabolite found in blood and is supplied as a high-purity research standard for metabolomics, lipidomics, and biomarker studies, including pregnancy and sepsis research.
  • High-purity reference standard (99.0%).
  • Molecular formula C25H52NO7P; molecular weight 509.66.
  • Solid powder form for storage and handling.
  • Stable as a powder at -20°C for up to 3 years.
  • Suitable for metabolomics and lipidomics assays.
  • Commonly used as an endogenous biomarker in pregnancy and sepsis research.