Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Heptadecene 99.5+%, TCI America™
SDP

Catalog No. S03475ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03475ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03475ML Supplier TCI America Supplier No. S03475ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6765-39-5
Molecular Formula C17H34
Molecular Weight (g/mol) 238.46
MDL Number MFCD00009000
InChI Key ADOBXTDBFNCOBN-UHFFFAOYSA-N
Synonym 1-heptadecene, heptadecene, hexahydroaplotaxene, alkenes, c>10 alpha, heptadecen, alkenes, c>10 .alpha., alpha-olefin wax, alpha olefins petroleum , c10 cut, c>10 alpha-alkenes, heptadecene related
PubChem CID 23217
ChEBI CHEBI:64502
IUPAC Name heptadec-1-ene
SMILES CCCCCCCCCCCCCCCC=C

Specifications

Color Yellow
Boiling Point 300°C
Quantity 5 mL
Formula Weight 238.46
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material 1-Heptadecene [Standard Material for GC]
EINECSNumber (2)-0027
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.