Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Heptafluorobutyryl)imidazole, 97%

Catalog No. AAL0022106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0022106 5 g
AAL0022114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0022106 Supplier Thermo Scientific Chemicals Supplier No. L0022106
Add to Cart

CAS: 32477-35-3 | C7H3F7N2O | 264.103 g/mol

1-(Heptafluorobutyryl)imidazole is used in gas chromatography for the determination of various pharmaceutical compounds like retronecine in biological matrixes. It acts as a reagent for derivatize amine-groups. It plays an essential role to reduce GC column degradation due to its non-acidic property. It is an acylating agent, which converts thermally unstable compounds to its more volatile derivative and then made suitable for GC analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Heptafluorobutyryl)imidazole is used in gas chromatography for the determination of various pharmaceutical compounds like retronecine in biological matrixes. It acts as a reagent for derivatize amine-groups. It plays an essential role to reduce GC column degradation due to its non-acidic property. It is an acylating agent, which converts thermally unstable compounds to its more volatile derivative and then made suitable for GC analysis.

Solubility
Miscible with chloroform and methanol.

Notes
Hygroscopic and air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32477-35-3
Molecular Formula C7H3F7N2O
Molecular Weight (g/mol) 264.103
MDL Number MFCD00014503
InChI Key MSYHGYDAVLDKCE-UHFFFAOYSA-N
Synonym n-heptafluorobutyrylimidazole, 1-heptafluorobutyryl imidazole, heptafluorobutyrylimidazole, n-heptafluorobutyryl imidazole, hfbi, 1-perfluorobutyryl imidazole, 2,2,3,3,4,4,4-heptafluoro-1-1h-imidazol-1-yl butan-1-one, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl-1h-imidazole, 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-yl butan-1-one, 1h-imidazole, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl
PubChem CID 94431
IUPAC Name 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-ylbutan-1-one
SMILES C1=CN(C=N1)C(=O)C(C(C(F)(F)F)(F)F)(F)F

Specifications

Melting Point 10°C to 13°C
Density 1.49
Boiling Point 160°C to 162°C
Flash Point 77°C (170°F)
Refractive Index 1.385
Quantity 5 g
Beilstein 4488026
Sensitivity Moisture sensitive
Solubility Information Miscible with chloroform and methanol.
Formula Weight 264.1
Percent Purity ≥98%
Chemical Name or Material 1-(Heptafluorobutyryl)imidazole
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 251-063-8
TSCA No
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.