Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Heptanethiol (Tech.), 90%, Thermo Scientific™

Catalog No. AC120410250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC120410250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC120410250 Supplier Thermo Scientific Chemicals Supplier No. 120410250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1639-09-4
CAS Min % 88.0
CAS Max % 100.0
Density 0.86g/mL
Infrared Spectrum Authentic
Assay Percent Range 90%
Molecular Formula C7H16S
Refractive Index 1.4480 to 1.4530 (20°C, 589nm)
Linear Formula CH3(CH2)6SH
Quantity 25g
Beilstein 01, 415
Synonym 1-heptanethiol, heptyl mercaptan, n-heptylmercaptan, heptyl thiol, heptylthiol, 1-heptylthiol, n-heptyl mercaptan, usaf ek-2122, unii-380ze61gr2, normal-heptyl mercaptan
InChI Key VPIAKHNXCOTPAY-UHFFFAOYSA-N
SMILES CCCCCCCS
IUPAC Name heptane-1-thiol
Molecular Weight (g/mol) 132.265
PubChem CID 15422
Formula Weight 132.26
Percent Purity ≥88%
Grade Technical
Chemical Name or Material 1-Heptanethiol, tech., 90%
Show More Show Less
EINECSNumber 216-678-8
RTECSNumber MJ1400000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.