Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Heptene, Spectrum™ Chemical
SDP

Catalog No. 18620875 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-875 100 mL
18-620-874 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-875 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H2666100ML
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synonym: Hept-1-ene, Heptylene, n-Hept-1-ene

Hept-1-ene, Heptylene, n-Hept-1-ene

Chemical Identifiers

CAS 592-76-7

Specifications

Boiling Point 94°C/201°F
Quantity 100 mL
Molecular Weight (g/mol) 98.19
Percent Purity 98%
Grade Ungraded
Physical Form Liquid
Chemical Name or Material 1-Heptene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.